Assay Design



Gene Transcripts

Gene Symbol
MTHFS
Entrez Gene ID
513372
Gene Name
5,10-methenyltetrahydrofolate synthetase (5-formyltetrahydrofolate cyclo-ligase)
Gene Aliases
-
Chromosome Location
Chr.21: 25950192 - 25967469 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075616.1 NP_001069084.1 2 - 3 446 69
GenBank mRNA BC114726.1 - 2 - 3 446 69

View Full Product Details