Assay Design



Gene Transcripts

Gene Symbol
ASB5
Entrez Gene ID
516722
Gene Name
ankyrin repeat and SOCS box containing 5
Gene Aliases
-
Chromosome Location
Chr.27: 7720262 - 7764695 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075744.1 NP_001069212.1 2 - 3 367 69
GenBank mRNA BC118205.1 - 2 - 3 367 69

View Full Product Details