Assay Design



Gene Transcripts

Gene Symbol
NUDCD3
Entrez Gene ID
533678
Gene Name
NudC domain containing 3
Gene Aliases
-
Chromosome Location
Chr.4: 76917158 - 76988089 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075997.1 NP_001069465.1 2 - 3 537 75
GenBank mRNA BC114136.1 - 2 - 3 537 75
BT021830.1 - 2 - 3 555 75

View Full Product Details