Assay Design



Gene Transcripts

Gene Symbol
NUDCD3
Entrez Gene ID
533678
Gene Name
NudC domain containing 3
Gene Aliases
-
Chromosome Location
Chr.4: 76917158 - 76988089 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075997.1 NP_001069465.1 3 - 4 663 99
GenBank mRNA BC114136.1 - 3 - 4 663 99
BT021830.1 - 3 - 4 681 99

View Full Product Details