Assay Design
Gene Transcripts
- Gene Symbol
- SLC25A33
- Entrez Gene ID
-
533794
- Gene Name
- solute carrier family 25 member 33
- Gene Aliases
-
-
- Chromosome Location
-
Chr.16: 43916562 - 43946759 on Build ARS-UCD1.2
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC116108.1 |
- |
3 - 4 |
504 |
101 |
View Full Product Details