Assay Design



Gene Transcripts

Gene Symbol
CIDEC
Entrez Gene ID
534607
Gene Name
cell death inducing DFFA like effector c
Gene Aliases
-
Chromosome Location
Chr.22: 16864388 - 16873816 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076031.1 NP_001069499.1 5 - 6 684 95
GenBank mRNA BC116135.1 - 5 - 6 684 95

View Full Product Details