Assay Design



Gene Transcripts

Gene Symbol
TMEFF2
Entrez Gene ID
538354
Gene Name
transmembrane protein with EGF like and two follistatin like domains 2
Gene Aliases
-
Chromosome Location
Chr.2: 80621359 - 80899341 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076101.2 NP_001069569.1 8 - 9 1283 121
GenBank mRNA BC118420.1 - 8 - 9 1284 121

View Full Product Details