Assay Design



Gene Transcripts

Gene Symbol
BOD1
Entrez Gene ID
540063
Gene Name
biorientation of chromosomes in cell division 1
Gene Aliases
FAM44B
Chromosome Location
Chr.20: 5319516 - 5331743 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076200.1 NP_001069668.1 2 - 3 507 92
GenBank mRNA BC114807.1 - 2 - 3 507 92

View Full Product Details