Assay Design



Gene Transcripts

Gene Symbol
EXTL2
Entrez Gene ID
540527
Gene Name
exostosin like glycosyltransferase 2
Gene Aliases
-
Chromosome Location
Chr.3: 42410212 - 42428919 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076224.1 NP_001069692.1 1 - 2 219 118
GenBank mRNA BC114730.1 - 1 - 2 219 118

View Full Product Details