Assay Design



Gene Transcripts

Gene Symbol
GDAP1L1
Entrez Gene ID
541158
Gene Name
ganglioside induced differentiation associated protein 1 like 1
Gene Aliases
-
Chromosome Location
Chr.13: 72715814 - 72740133 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076261.1 NP_001069729.1 3 - 4 638 64
GenBank mRNA BC133446.1 - 3 - 4 568 64
BT025400.1 - 3 - 4 638 64

View Full Product Details