Assay Design



Gene Transcripts

Gene Symbol
RAB8B
Entrez Gene ID
614242
Gene Name
RAB8B, member RAS oncogene family
Gene Aliases
-
Chromosome Location
Chr.10: 46780496 - 46850532 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076317.1 NP_001069785.1 4 - 5 339 121
GenBank mRNA BC105330.1 - 4 - 5 339 121

View Full Product Details