Assay Design



Gene Transcripts

Gene Symbol
LMO2
Entrez Gene ID
614876
Gene Name
LIM domain only 2
Gene Aliases
-
Chromosome Location
Chr.15: 64472394 - 64505635 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076352.1 NP_001069820.1 1 - 2 256 58
GenBank mRNA BC116134.1 - 1 - 2 256 58

View Full Product Details