Assay Design



Gene Transcripts

Gene Symbol
MGST2
Entrez Gene ID
615552
Gene Name
microsomal glutathione S-transferase 2
Gene Aliases
-
Chromosome Location
Chr.17: 18024427 - 18062298 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076382.1 NP_001069850.1 2 - 3 267 89
GenBank mRNA BC105356.1 - 2 - 3 235 89
BT025439.1 - 2 - 3 267 89

View Full Product Details