Assay Design



Gene Transcripts

Gene Symbol
COA8
Entrez Gene ID
617441
Gene Name
cytochrome c oxidase assembly factor 8
Gene Aliases
APOP-1, APOPT1, C14orf153, C21H14orf153
Chromosome Location
Chr.21: 68196002 - 68225883 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076459.2 NP_001069927.1 1 - 2 150 113
GenBank mRNA BC118424.1 - 1 - 2 152 113
BT025435.1 - 1 - 2 124 113

View Full Product Details