Assay Design



Gene Transcripts

Gene Symbol
HAUS2
Entrez Gene ID
508698
Gene Name
HAUS augmin like complex subunit 2
Gene Aliases
CEP27
Chromosome Location
Chr.10: 37875094 - 37888395 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076819.2 NP_001070287.1 2 - 3 203 103
GenBank mRNA BC105449.1 - 2 - 3 208 103
BT030707.1 - 2 - 3 203 103

View Full Product Details