Assay Design



Gene Transcripts

Gene Symbol
MS4A1
Entrez Gene ID
505653
Gene Name
membrane spanning 4-domains A1
Gene Aliases
CD20
Chromosome Location
Chr.15: 83282895 - 83298116 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077854.2 NP_001071322.1 6 - 7 854 126
GenBank mRNA BC118132.1 - 6 - 7 855 126

View Full Product Details