Assay Design



Gene Transcripts

Gene Symbol
ESAM
Entrez Gene ID
616926
Gene Name
endothelial cell adhesion molecule
Gene Aliases
-
Chromosome Location
Chr.29: 28232745 - 28241470 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001078066.2 NP_001071534.1 5 - 6 895 58
GenBank mRNA BC105434.1 - 5 - 6 896 58

View Full Product Details