Assay Design



Gene Transcripts

Gene Symbol
GPATCH4
Entrez Gene ID
768314
Gene Name
G-patch domain containing 4
Gene Aliases
GPATC4
Chromosome Location
Chr.3: 14232874 - 14240974 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001078129.2 NP_001071597.1 1 - 2 29 64
GenBank mRNA BC105388.1 - 1 - 2 30 64

View Full Product Details