Assay Design



Gene Transcripts

Gene Symbol
JKAMP
Entrez Gene ID
509181
Gene Name
JNK1/MAPK8 associated membrane protein
Gene Aliases
JAMP
Chromosome Location
Chr.10: 71759901 - 71782137 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080247.1 NP_001073716.1 3 - 4 259 112
GenBank mRNA BC116146.1 - 3 - 4 259 112
BC146270.1 - 3 - 4 295 112
BT025457.1 - 2 - 3 259 112

View Full Product Details