Assay Design



Gene Transcripts

Gene Symbol
TUT1
Entrez Gene ID
616339
Gene Name
terminal uridylyl transferase 1, U6 snRNA-specific
Gene Aliases
RBM21
Chromosome Location
Chr.29: 40927674 - 40937085 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080322.1 NP_001073791.1 1 - 2 116 56
GenBank mRNA BT025417.1 - 1 - 2 116 56

View Full Product Details