Assay Design



Gene Transcripts

Gene Symbol
RAB3A
Entrez Gene ID
282029
Gene Name
RAB3A, member RAS oncogene family
Gene Aliases
-
Chromosome Location
Chr.7: 5018104 - 5023793 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174446.3 NP_776871.2 2 - 3 339 83
GenBank mRNA BC118278.1 - 2 - 3 339 83
M19885.1 - 2 - 3 275 83

View Full Product Details