Assay Design
Gene Transcripts
- Gene Symbol
- RAB3A
- Entrez Gene ID
-
282029
- Gene Name
- RAB3A, member RAS oncogene family
- Gene Aliases
-
-
- Chromosome Location
-
Chr.7: 5018104 - 5023793 on Build ARS-UCD1.2
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_174446.3 |
NP_776871.2 |
3 - 4 |
456 |
65 |
GenBank mRNA |
BC118278.1 |
- |
3 - 4 |
456 |
65 |
View Full Product Details