Assay Design



Gene Transcripts

Gene Symbol
UPK1B
Entrez Gene ID
282113
Gene Name
uroplakin 1B
Gene Aliases
-
Chromosome Location
Chr.1: 63981479 - 64010646 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174482.2 NP_776907.2 7 - 8 858 69
GenBank mRNA BC116014.1 - 7 - 8 858 69
Z29378.1 - - 790 69

View Full Product Details