Assay Design



Gene Transcripts

Gene Symbol
P2RY12
Entrez Gene ID
408007
Gene Name
purinergic receptor P2Y12
Gene Aliases
-
Chromosome Location
Chr.1: 116694848 - 116747195 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001174.2 NP_001001174.2 1 - 2 53 61
GenBank mRNA BC122591.1 - 1 - 2 53 61

View Full Product Details