Assay Design



Gene Transcripts

Gene Symbol
SLC25A38
Entrez Gene ID
512325
Gene Name
solute carrier family 25 member 38
Gene Aliases
-
Chromosome Location
Chr.22: 12645740 - 12660768 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014905.2 NP_001014905.2 4 - 5 701 72
GenBank mRNA BC148053.1 - 4 - 5 725 72
BT020649.1 - 4 - 5 740 72
BT020880.1 - 4 - 5 728 72
BT021801.1 - 4 - 5 699 72
BT026157.1 - 4 - 5 701 72

View Full Product Details