Assay Design



Gene Transcripts

Gene Symbol
GCAT
Entrez Gene ID
319141
Gene Name
glycine C-acetyltransferase
Gene Aliases
-
Chromosome Location
Chr.5: 109622546 - 109629294 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075134.1 NP_001068602.1 7 - 8 990 64
GenBank mRNA BC119870.1 - 7 - 8 990 64

View Full Product Details