Assay Design



Gene Transcripts

Gene Symbol
FMO2
Entrez Gene ID
504401
Gene Name
flavin containing dimethylaniline monoxygenase 2
Gene Aliases
-
Chromosome Location
Chr.16: 38756046 - 38800243 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075162.2 NP_001068630.1 4 - 5 613 80
NM_001163274.3 NP_001156746.2 4 - 5 613 80
GenBank mRNA BC120448.1 - 4 - 5 533 80
EH159834.1 - 4 - 5 579 80

View Full Product Details