Assay Design



Gene Transcripts

Gene Symbol
SNF8
Entrez Gene ID
505628
Gene Name
SNF8 subunit of ESCRT-II
Gene Aliases
-
Chromosome Location
Chr.19: 37587438 - 37596408 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075211.1 NP_001068679.1 7 - 8 713 150
GenBank mRNA BC123598.1 - 7 - 8 713 150

View Full Product Details