Assay Design



Gene Transcripts

Gene Symbol
WDR45B
Entrez Gene ID
506232
Gene Name
WD repeat domain 45B
Gene Aliases
WDR45L
Chromosome Location
Chr.19: 49991652 - 50015615 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075247.1 NP_001068715.1 9 - 10 993 93
GenBank mRNA BC119890.1 - 9 - 10 993 93

View Full Product Details