Assay Design



Gene Transcripts

Gene Symbol
AURKAIP1
Entrez Gene ID
506359
Gene Name
aurora kinase A interacting protein 1
Gene Aliases
-
Chromosome Location
Chr.16: 51147423 - 51151897 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075255.1 NP_001068723.1 2 - 3 118 65
GenBank mRNA BC120143.1 - 2 - 3 118 65

View Full Product Details