Assay Design



Gene Transcripts

Gene Symbol
ATF1
Entrez Gene ID
506967
Gene Name
activating transcription factor 1
Gene Aliases
-
Chromosome Location
Chr.5: 29107516 - 29171845 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075289.1 NP_001068757.1 6 - 7 934 135
GenBank mRNA BC123855.1 - 6 - 7 934 135

View Full Product Details