Assay Design



Gene Transcripts

Gene Symbol
MYOT
Entrez Gene ID
507884
Gene Name
myotilin
Gene Aliases
-
Chromosome Location
Chr.7: 49339573 - 49357206 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075335.1 NP_001068803.1 9 - 10 1594 64
GenBank mRNA BC119939.1 - 9 - 10 1594 64

View Full Product Details