Assay Design



Gene Transcripts

Gene Symbol
MFSD5
Entrez Gene ID
507921
Gene Name
major facilitator superfamily domain containing 5
Gene Aliases
-
Chromosome Location
Chr.5: 26756799 - 26760156 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075340.1 NP_001068808.1 1 - 2 95 66
GenBank mRNA BC120417.1 - 1 - 2 95 66

View Full Product Details