Assay Design



Gene Transcripts

Gene Symbol
TP53I3
Entrez Gene ID
508875
Gene Name
tumor protein p53 inducible protein 3
Gene Aliases
-
Chromosome Location
Chr.11: 74983273 - 74989608 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075378.1 NP_001068846.1 4 - 5 929 139
GenBank mRNA BC120317.1 - 4 - 5 929 139

View Full Product Details