Assay Design



Gene Transcripts

Gene Symbol
RARRES1
Entrez Gene ID
510102
Gene Name
retinoic acid receptor responder 1
Gene Aliases
-
Chromosome Location
Chr.1: 108807575 - 108849587 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075430.2 NP_001068898.1 2 - 3 341 104
GenBank mRNA BC120187.1 - 2 - 3 343 104

View Full Product Details