Assay Design



Gene Transcripts

Gene Symbol
CCDC174
Entrez Gene ID
512697
Gene Name
coiled-coil domain containing 174
Gene Aliases
C22H3orf19
Chromosome Location
Chr.22: 57604754 - 57628100 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075578.1 NP_001069046.1 8 - 9 826 142
GenBank mRNA BC119912.1 - 8 - 9 826 142

View Full Product Details