Assay Design



Gene Transcripts

Gene Symbol
PCYOX1L
Entrez Gene ID
514598
Gene Name
prenylcysteine oxidase 1 like
Gene Aliases
-
Chromosome Location
Chr.7: 60755244 - 60767538 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075673.1 NP_001069141.1 5 - 6 834 70
GenBank mRNA BC120048.1 - 5 - 6 834 70

View Full Product Details