Assay Design



Gene Transcripts

Gene Symbol
PAXBP1
Entrez Gene ID
514918
Gene Name
PAX3 and PAX7 binding protein 1
Gene Aliases
GCFC, GCFC1
Chromosome Location
Chr.1: 2786812 - 2819637 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075689.3 NP_001069157.3 12 - 13 2077 100
GenBank mRNA BC120358.1 - 6 - 7 1016 100

View Full Product Details