Assay Design



Gene Transcripts

Gene Symbol
NFKBIB
Entrez Gene ID
525408
Gene Name
NFKB inhibitor beta
Gene Aliases
-
Chromosome Location
Chr.18: 48614347 - 48624970 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075872.1 NP_001069340.1 4 - 5 770 106
GenBank mRNA BC122726.1 - 4 - 5 770 106

View Full Product Details