Assay Design



Gene Transcripts

Gene Symbol
KLK10
Entrez Gene ID
526736
Gene Name
kallikrein related peptidase 10
Gene Aliases
-
Chromosome Location
Chr.18: 56989035 - 56995323 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075890.1 NP_001069358.1 1 - 2 65 85
GenBank mRNA BC119919.1 - 1 - 2 65 85

View Full Product Details