Assay Design



Gene Transcripts

Gene Symbol
CALY
Entrez Gene ID
531593
Gene Name
calcyon neuron specific vesicular protein
Gene Aliases
-
Chromosome Location
Chr.26: 25666546 - 25677469 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075953.2 NP_001069421.1 5 - 6 781 140
GenBank mRNA BC120132.1 - 5 - 6 782 140

View Full Product Details