Assay Design



Gene Transcripts

Gene Symbol
INA
Entrez Gene ID
532236
Gene Name
internexin neuronal intermediate filament protein alpha
Gene Aliases
-
Chromosome Location
Chr.26: 23887978 - 23900124 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075958.1 NP_001069426.1 1 - 2 1116 66
GenBank mRNA BC123743.1 - 1 - 2 1116 66

View Full Product Details