Assay Design
Gene Transcripts
- Gene Symbol
- COQ5
- Entrez Gene ID
-
533208
- Gene Name
- coenzyme Q5, methyltransferase
- Gene Aliases
-
-
- Chromosome Location
-
Chr.17: 62796218 - 62813415 on Build ARS-UCD1.2
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC120308.1 |
- |
5 - 6 |
776 |
104 |
View Full Product Details