Assay Design



Gene Transcripts

Gene Symbol
HEBP1
Entrez Gene ID
533227
Gene Name
heme binding protein 1
Gene Aliases
-
Chromosome Location
Chr.5: 96875437 - 96912825 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075985.2 NP_001069453.1 2 - 3 249 105
GenBank mRNA BC118459.1 - 2 - 3 250 105

View Full Product Details