Assay Design



Gene Transcripts

Gene Symbol
FBXO25
Entrez Gene ID
540328
Gene Name
F-box protein 25
Gene Aliases
-
Chromosome Location
Chr.1: 146528087 - 146556467 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076211.1 NP_001069679.1 5 - 6 497 67
GenBank mRNA BC114733.1 - 5 - 6 497 67

View Full Product Details