Assay Design



Gene Transcripts

Gene Symbol
SRSF1
Entrez Gene ID
615796
Gene Name
serine and arginine rich splicing factor 1
Gene Aliases
SFRS1
Chromosome Location
Chr.19: 8806072 - 8810092 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076394.1 NP_001069862.1 3 - 4 648 118
GenBank mRNA BC119930.1 - 3 - 4 648 118

View Full Product Details