Assay Design



Gene Transcripts

Gene Symbol
ERGIC1
Entrez Gene ID
616293
Gene Name
endoplasmic reticulum-golgi intermediate compartment 1
Gene Aliases
-
Chromosome Location
Chr.20: 4596537 - 4709462 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076417.1 NP_001069885.1 8 - 9 741 97
GenBank mRNA BC120357.1 - 8 - 9 741 97

View Full Product Details