Assay Design



Gene Transcripts

Gene Symbol
FARS2
Entrez Gene ID
505390
Gene Name
phenylalanyl-tRNA synthetase 2, mitochondrial
Gene Aliases
-
Chromosome Location
Chr.23: 49111514 - 49417402 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076805.1 NP_001070273.1 6 - 7 1230 62
GenBank mRNA BC123887.1 - 6 - 7 1230 62

View Full Product Details