Assay Design



Gene Transcripts

Gene Symbol
COL3A1
Entrez Gene ID
510833
Gene Name
collagen type III alpha 1 chain
Gene Aliases
-
Chromosome Location
Chr.2: 7375278 - 7414912 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076831.1 NP_001070299.1 48 - 49 3902 70
GenBank mRNA BC123469.1 - 48 - 49 3902 70

View Full Product Details