Assay Design



Gene Transcripts

Gene Symbol
HNRNPLL
Entrez Gene ID
534315
Gene Name
heterogeneous nuclear ribonucleoprotein L like
Gene Aliases
HNRPLL
Chromosome Location
Chr.11: 20909750 - 20948162 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076900.1 NP_001070368.1 2 - 3 556 72
GenBank mRNA BC123451.1 - 2 - 3 556 72

View Full Product Details